[4-(4-Chlorophenyl)phenyl]sulfonyl chloride 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4'-Chloro-[1,1'-biphenyl]-4-sulphonyl chloride, 4'-Chlorobiphenyl-4-sulphonyl chloride

H34182-03 H34182-09
AAH34182-03EA 46 USD
AAH34182-03 AAH34182-09
[4-(4-Chlorophenyl)phenyl]sulfonyl chloride 96%
[4-(4-Chlorophenyl)phenyl]sulfonyl chloride

Purity: 96% CAS No.: 20443-74-7 Molecular Formula: C12H8Cl2O2S Formula Weight: 287.17 Melting Point: 107-108° UN No.: 3261 MDL No.: MFCD01631918

Formula: C₁₂H₈Cl₂O₂S
MW: 287.17 g/mol
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD01631918
CAS Number: 20443-74-7
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR