1-Methylindole-3-carbonitrile 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-1H-indole-3-carbonitrile

H34157-03 H34157-06 H34157-MD
AAH34157-03EA 44.3 USD
AAH34157-03 AAH34157-06 AAH34157-MD
1-Methylindole-3-carbonitrile 96%
1-Methylindole-3-carbonitrile

Purity: 96% CAS No.: 24662-37-1 Molecular Formula: C10H8N2 Formula Weight: 156.19 Melting Point: 55-56° MDL No.: MFCD00466602

Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Melting Pt: 55…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00466602
CAS Number: 24662-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR