2-Amino-4-chloro-3,5,6-trifluoropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34155-03
H34155-06
AAH34155-03EA
86.7
USD
AAH34155-03
AAH34155-06
AAH34155-MD
2-Amino-4-chloro-3,5,6-trifluoropyridine 98%
2-Amino-4-chloro-3,5,6-trifluoropyridine
Purity: 98%
CAS No.: 63489-56-5
Molecular Formula: C5H2ClF3N2
Formula Weight: 182.54
Melting Point: 119-123°
MDL No.: MFCD08436088
Formula:
C₅H₂ClF₃N₂ MW: 182.54 g/mol Melting Pt: 119…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD08436088 CAS Number: 63489-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...