3-(2-Fluorophenyl)propionic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

H34110-03 H34110-09
AAH34110-03EA 37.5 USD
AAH34110-03 AAH34110-09
3-(2-Fluorophenyl)propionic acid 96%
3-(2-Fluorophenyl)propionic acid

Purity: 96% CAS No.: 1643-26-1 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Melting Point: 75-79° MDL No.: MFCD01310820

Formula: C₉H₉FO₂
MW: 168.17 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD01310820
CAS Number: 1643-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR