2-Fluoro-4-(pentafluorothio)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H34099-MD H34099-03
AAH34099-MDEA 249 USD
AAH34099-MD AAH34099-03
2-Fluoro-4-(pentafluorothio)aniline 97%
2-Fluoro-4-(pentafluorothio)aniline

Purity: 97% CAS No.: 1240257-25-3 Molecular Formula: C6H5F6NS Formula Weight: 237.16 UN No.: 2810 MDL No.: MFCD16652511

Formula: C₆H₅F₆NS
MW: 237.16
Storage Temperature: Ambient
MDL Number: MFCD16652511
CAS Number: 1240257-25-3
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR