Lithium-4,5-dicyano-2-(trifluoromethyl)imidazole 95%

Supplier: Thermo Scientific Chemicals
Warning

H34081-03 H34081-09
AAH34081-03EA 97.8 USD
AAH34081-03 AAH34081-09
Lithium-4,5-dicyano-2-(trifluoromethyl)imidazole 95%
Lithium-4,5-dicyano-2-(trifluoromethyl)imidazole

Purity: 95% Molecular Formula: C6F3LiN4 Formula Weight: 192.03 MDL No.: MFCD18827480

Formula: C₆F₃LiN₄
MW: 192.03 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD18827480
EINECS: 691-963-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR