5-Amino-2-methylbenzenesulfonamide 96%

Supplier: Thermo Scientific Chemicals
Warning

H34056-03 H34056-06
AAH34056-03EA 28.6 USD
AAH34056-03 AAH34056-06
5-Amino-2-methylbenzenesulfonamide 96%
5-Amino-2-methylbenzenesulphonamide

Purity: 96% CAS No.: 6973-09-7 Molecular Formula: C7H10N2O2S Formula Weight: 186.23 Melting Point: 163-164° MDL No.: MFCD06681071

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Melting Pt: 163…164 °C
Storage Temperature: Ambient
MDL Number: MFCD06681071
CAS Number: 6973-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR