Ethyl-6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

H33992-03 H33992-06
AAH33992-03EA 80.9 USD
AAH33992-03 AAH33992-06 AAH33992-MD
Ethyl-6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 98%
Ethyl-6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

Purity: 98% CAS No.: 1027511-41-6 Molecular Formula: C9H8BrN3O2 Formula Weight: 270.08 Melting Point: 144-146° MDL No.: MFCD10574908

Formula: C₉H₈BrN₃O₂
MW: 270.08 g/mol
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD10574908
CAS Number: 1027511-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR