Methyl-3-aminobenzo[b]thiophene-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

H33956-03 H33956-06
AAH33956-03EA 40.9 USD
AAH33956-03 AAH33956-06
Methyl-3-aminobenzo[b]thiophene-2-carboxylate 97%
Methyl 3-aminobenzo[b]thiophene-2-carboxylate

Purity: 97% CAS No.: 35212-85-2 Molecular Formula: C10H9NO2S Formula Weight: 207.25 Melting Point: 107-108° MDL No.: MFCD00206744

Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00206744
CAS Number: 35212-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR