4-Methylbenzoic anhydride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-methylbenzene-1-carboxylic anhydride
H33951-03
H33951-06
AAH33951-03EA
38.1
USD
AAH33951-03
AAH33951-06
4-Methylbenzoic anhydride 97%
4-Methylbenzoic anhydride
Purity: 97%
CAS No.: 13222-85-0
Molecular Formula: C16H14O3
Formula Weight: 254.29
Melting Point: 86-89°
MDL No.: MFCD00048100
Formula:
C₁₆H₁₄O₃ Melting Pt: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00048100 CAS Number: 13222-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...