3-[3-(Benzyloxy)phenyl]propionic acid 96%
Supplier: Thermo Scientific Chemicals
H33899-03
H33899-06
AAH33899-03EA
98.8
USD
AAH33899-03
AAH33899-06
AAH33899-MD
3-[3-(Benzyloxy)phenyl]propionic acid 96%
3-[3-(Benzyloxy)phenyl]propionic acid
Purity: 96%
CAS No.: 57668-34-5
Molecular Formula: C16H16O3
Formula Weight: 256.30
Melting Point: 83-85°
MDL No.: MFCD09258905
Formula:
C₁₆H₁₆O₃ MW: 256.30 g/mol Melting Pt: 83…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258905 CAS Number: 57668-34-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...