6-Bromopyrazolo[1,5-a]pyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

H33855-03 H33855-06
AAH33855-03EA 96.9 USD
AAH33855-03 AAH33855-06
6-Bromopyrazolo[1,5-a]pyrimidine 97%
6-Bromopyrazolo[1,5-a]pyrimidine

Purity: 97% CAS No.: 705263-10-1 Molecular Formula: C6H4BrN3 Formula Weight: 198.02 Melting Point: 122-124° MDL No.: MFCD09832894

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09832894
CAS Number: 705263-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR