1-Chloromethyl-1H-1,2,4-triazole hydrochloride 96%

Supplier: Thermo Scientific Chemicals
Warning

H33830-03 H33830-06
AAH33830-03EA 101.32 USD
AAH33830-03 AAH33830-06 AAH33830-MD
1-Chloromethyl-1H-1,2,4-triazole hydrochloride 96%
1-Chloromethyl-1H-1,2,4-triazole hydrochloride

Purity: 96% CAS No.: 84387-62-2 Molecular Formula: C3H4ClN3.HCl Formula Weight: 154.00 Melting Point: 138-145° MDL No.: MFCD00964947

Formula: C₃H₄ClN₃·HCl
Melting Pt: 138…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00964947
CAS Number: 84387-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR