2-Methylbenzothiazol-6-ol 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylbenzo[d]thiazol-6-ol

H33812-03 H33812-06
AAH33812-03EA 105 USD
AAH33812-03 AAH33812-06 AAH33812-MD
2-Methylbenzothiazol-6-ol 96%
2-Methylbenzothiazol-6-ol

Purity: 96% CAS No.: 68867-18-5 Molecular Formula: C8H7NOS Formula Weight: 165.21 Melting Point: 159-161° MDL No.: MFCD00227234

Formula: C₈H₇NOS
MW: 165.21 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00227234
CAS Number: 68867-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR