4-(Trifluoromethyl)cyclohexanone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33793-03
H33793-09
AAH33793-03EA
104
USD
AAH33793-03
AAH33793-09
4-(Trifluoromethyl)cyclohexanone 97%
4-(Trifluoromethyl)cyclohexanone
Purity: 97%
CAS No.: 75091-99-5
Molecular Formula: C7H9F3O
Formula Weight: 166.14
Boiling Point: 67-68°/16mm
UN No.: 1224
MDL No.: MFCD00102145
Formula:
C₇H₉F₃O MW: 166.14 Boiling Pt: 67…68 °C/16 mm Storage Temperature: Ambient |
MDL Number:
MFCD00102145 CAS Number: 75091-99-5 UN: 1224 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...