3-(Trifluoromethyl)benzoic anhydride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33779-03
H33779-06
AAH33779-03EA
32.4
USD
AAH33779-03
AAH33779-06
3-(Trifluoromethyl)benzoic anhydride 97%
3-(Trifluoromethyl)benzoic anhydride
Purity: 97%
CAS No.: 25753-15-5
Molecular Formula: C16H8F6O3
Formula Weight: 362.23
Melting Point: 74-78°
MDL No.: MFCD09757594
Formula:
C₁₆H₈F₆O₃ MW: 362.23 g/mol Melting Pt: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD09757594 CAS Number: 25753-15-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...