3-(4-Benzyloxyphenyl)propionic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33766-03
H33766-06
AAH33766-03EA
26.7
USD
AAH33766-03
AAH33766-06
3-(4-Benzyloxyphenyl)propionic acid 96%
3-(4-Benzyloxyphenyl)propionic acid
Purity: 96%
CAS No.: 50463-48-4
Molecular Formula: C16H16O3
Formula Weight: 256.30
Melting Point: 116-118°
MDL No.: MFCD01570264
Formula:
C₁₆H₁₆O₃ Melting Pt: 116…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD01570264 CAS Number: 50463-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...