Ethyl-3-cyclopropyl-3-oxopropanoate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-cyclopropyl-3-oxopropanoate

H33756-03 H33756-06
AAH33756-03EA 87.5 USD
AAH33756-03 AAH33756-06
Ethyl-3-cyclopropyl-3-oxopropanoate 96%
Ethyl-3-cyclopropyl-3-oxopropanoate

Purity: 96% CAS No.: 24922-02-9 Molecular Formula: C8H12O3 Formula Weight: 156.18 Boiling Point: 73°/1mm MDL No.: MFCD00184570

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Boiling Pt: 73 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD00184570
CAS Number: 24922-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR