2-Bromo-4-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H33698-03 H33698-06
AAH33698-03EA 39.4 USD
AAH33698-03 AAH33698-06
2-Bromo-4-chloropyridine 97%
2-Bromo-4-chloropyridine

Purity: 97% CAS No.: 22918-01-0 Molecular Formula: C5H3BrClN Formula Weight: 192.44 Melting Point: 30° MDL No.: MFCD03840757

Formula: C₅H₃BrClN
Melting Pt: Storage Temperature: Ambient
MDL Number: MFCD03840757
CAS Number: 22918-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR