3-Bromofuro[3,2-b]pyridine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33652-03
H33652-06
AAH33652-03EA
213
USD
AAH33652-03
AAH33652-06
AAH33652-MD
3-Bromofuro[3,2-b]pyridine 96%
3-Bromofuro[3,2-b]pyridine
Purity: 96%
CAS No.: 92404-63-2
Molecular Formula: C7H4BrNO
Formula Weight: 198.02
Storage Temperature: 2°C to 8°C
Melting Point: 51-53°
MDL No.: MFCD19687763
Formula:
C₇H₄BrNO MW: 198.02 g/mol Melting Pt: 51…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD19687763 CAS Number: 92404-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...