N-(4-Methyl-2-pyridyl)thiourea 97%

Supplier: Thermo Scientific Chemicals
Warning

H33648-03 H33648-06
AAH33648-03EA 50.4 USD
AAH33648-03 AAH33648-06
N-(4-Methyl-2-pyridyl)thiourea 97%
N-(4-Methyl-2-pyridyl)thiourea

Purity: 97% CAS No.: 21242-21-7 Molecular Formula: C7H9N3S Formula Weight: 167.23 Melting Point: 216-220° MDL No.: MFCD04117348

Formula: C₇H₉N₃S
Melting Pt: 216…220 °C
Storage Temperature: Ambient
MDL Number: MFCD04117348
CAS Number: 21242-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR