Rhodamine 6G perchlorate 96%

Supplier: Thermo Scientific Chemicals
Warning

H33626-03 H33626-MD
AAH33626-03EA 108 USD
AAH33626-03 AAH33626-MD
Rhodamine 6G perchlorate 96%
Rhodamine 6G perchlorate

Purity: 96% CAS No.: 13161-28-9 Molecular Formula: C28H31ClN2O7 Formula Weight: 543.02 Melting Point: 240° MDL No.: MFCD00013117

Formula: C₂₈H₃₁ClN₂O₇
MW: 543.02 g/mol
Melting Pt: >240 °C
Storage Temperature: Ambient
MDL Number: MFCD00013117
CAS Number: 13161-28-9
EINECS: 236-099-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR