3-(3-Chloro-4-fluorophenyl)propionic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33592-03
H33592-06
AAH33592-03EA
68.7
USD
AAH33592-03
AAH33592-06
3-(3-Chloro-4-fluorophenyl)propionic acid 96%
3-(3-Chloro-4-fluorophenyl)propionic acid
Purity: 96%
CAS No.: 881190-93-8
Molecular Formula: C9H8ClFO2
Formula Weight: 202.61
Melting Point: 56-59°
MDL No.: MFCD04116056
Formula:
C₉H₈ClFO₂ MW: 202.61 g/mol Melting Pt: 56…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD04116056 CAS Number: 881190-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...