3-(2-Chloro-6-fluorophenyl)propionic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2-Chloro-6-fluorophenyl)propanoic acid

H33586-03 H33586-06
AAH33586-03EA 92.5 USD
AAH33586-03 AAH33586-06
3-(2-Chloro-6-fluorophenyl)propionic acid 96%
3-(2-Chloro-6-fluorophenyl)propionic acid

Purity: 96% CAS No.: 88740-77-6 Molecular Formula: C9H8ClFO2 Formula Weight: 202.61 Melting Point: 59-63° MDL No.: MFCD06660331

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD06660331
CAS Number: 88740-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR