2,3-Dihydro-1-benzofuran-5-sulfonoylchloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33579-06
H33579-MD
AAH33579-06EA
112
USD
AAH33579-06
AAH33579-MD
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride 97%
2,3-Dihydro-1-benzofuran-5-sulfonoylchloride
Purity: 97%
CAS No.: 115010-11-2
Molecular Formula: C8H7ClO3S
Formula Weight: 218.66
Storage Temperature: 2°C to 8°C
Melting Point: 83-85°
UN No.: 3261
MDL No.: MFCD02677703
Formula:
C₈H₇ClO₃S MW: 218.66 g/mol Melting Pt: 83…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD02677703 CAS Number: 115010-11-2 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...