3-Amino-5-phenyl-1,2,4-thiadiazole 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33552-03
H33552-06
H33552-MD
AAH33552-03EA
115
USD
AAH33552-03
AAH33552-06
AAH33552-MD
3-Amino-5-phenyl-1,2,4-thiadiazole 96%
3-Amino-5-phenyl-1,2,4-thiadiazole
Purity: 96%
CAS No.: 27182-54-3
Molecular Formula: C8H7N3S
Formula Weight: 177.23
Melting Point: 136-137°
MDL No.: MFCD00159799
Formula:
C₈H₇N₃S Melting Pt: 136…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00159799 CAS Number: 27182-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...