5-Bromo-3-iodo-2-hydroxypyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33495-03
H33495-06
H33495-14
AAH33495-03EA
21.5
USD
AAH33495-03
AAH33495-06
AAH33495-14
5-Bromo-3-iodo-2-hydroxypyridine 97%
5-Bromo-3-iodo-2-hydroxypyridine
Purity: 97%
CAS No.: 381233-75-6
Molecular Formula: C5H3BrINO
Formula Weight: 299.89
Melting Point: 226-227°
MDL No.: MFCD08688586
Formula:
C₅H₃BrINO Melting Pt: 226…227 °C Storage Temperature: Ambient |
MDL Number:
MFCD08688586 CAS Number: 381233-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...