3,3,3-Trifluoro-DL-alanine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-3,3,3-trifluoropropanoic acid, H-DL-3,3,3-TriF-Ala-OH

H33461-03 H33461-MD
AAH33461-03EA 428 USD
AAH33461-03 AAH33461-MD
3,3,3-Trifluoro-DL-alanine 97%
3,3,3-Trifluoro-DL-alanine

Purity: 97% CAS No.: 17463-43-3 Molecular Formula: C3H4F3NO2 Formula Weight: 143.06 Melting Point: 210° dec. MDL No.: MFCD00004263

Formula: C₃H₄F₃NO₂
MW: 143.07 g/mol
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004263
CAS Number: 17463-43-3
EINECS: 241-478-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR