Methyl-2-chloro-6-methoxyisonicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-chloro-6-methoxypyridine-4-carboxylate, Methyl 2-chloro-6-methoxyisonicotinate

H33438-06 H33438-14
AAH33438-06EA 126 USD
AAH33438-06 AAH33438-14 AAH33438-03
Methyl-2-chloro-6-methoxyisonicotinate 98%
Methyl 2-chloro-6-methoxyisonicotinate

Purity: 98% CAS No.: 42521-10-8 Molecular Formula: C8H8ClNO3 Formula Weight: 201.61 Melting Point: 103-105° MDL No.: MFCD00173929

Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00173929
CAS Number: 42521-10-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR