Methyl-1-boc-3-pyrrolidinecarboxylate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Pyrrolidine-1,3-dicarboxylic acid 1-tert-Butyl 3-Methyl ester, 1-Boc-3-pyrrolidinecarboxylic acid methyll ester, 1-tert-Butyl 3-Methyl Pyrrolidine-1,3-dicarboxylate, 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid methyll ester, Methyl 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylate, Methyl 1-Boc-3-pyrrolidinecarboxylate
H33407-03
H33407-06
AAH33407-03EA
111
USD
AAH33407-03
AAH33407-06
Methyl-1-boc-3-pyrrolidinecarboxylate 97%
Methyl-1-boc-3-pyrrolidinecarboxylate
Purity: 97%
CAS No.: 122684-33-7
Molecular Formula: C11H19NO4
Formula Weight: 229.28
Boiling Point: 105°/0.2mm
MDL No.: MFCD04038683
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol Boiling Pt: 105 °C/0.2 mm Storage Temperature: Ambient |
MDL Number:
MFCD04038683 CAS Number: 122684-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...