1-(4-Methylpyridin-3-yl)ethanone 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Methyl-3-pyridinyl)-Ethanone

H33380-03 H33380-06
AAH33380-03EA 69.2 USD
AAH33380-03 AAH33380-06
1-(4-Methylpyridin-3-yl)ethanone 96%
1-(4-Methylpyridin-3-yl)ethanone

Purity: 96% CAS No.: 51227-30-6 Molecular Formula: C8H9NO Formula Weight: 135.17 Refractive Index: 1.5295 Boiling Point: 57-58°/1mm MDL No.: MFCD08447082

Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 57…58 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD08447082
CAS Number: 51227-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR