2-(Trifluoromethyl)pyrazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H33338-03 H33338-06
AAH33338-03EA 85.6 USD
AAH33338-03 AAH33338-06
2-(Trifluoromethyl)pyrazine 97%
2-(Trifluoromethyl)pyrazine

Purity: 97% CAS No.: 61655-67-2 Molecular Formula: C5H3F3N2 Formula Weight: 148.09 Boiling Point: 116-118° UN No.: 2810 MDL No.: MFCD06797734

Formula: C₅H₃F₃N₂
MW: 148.09 g/mol
Boiling Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD06797734
CAS Number: 61655-67-2
UN: 1993
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR