4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Trifluoromethyl)cyclohexanol, 4-(Trifluoromethyl)cyclohexan-1-ol

H33317-03 H33317-06
AAH33317-03EA 85.5 USD
AAH33317-03 AAH33317-06
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) 97%
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)

Purity: 97% CAS No.: 30129-18-1 Molecular Formula: C7H11F3O Formula Weight: 168.16 Boiling Point: 70-75°/15mm MDL No.: MFCD00102144

Formula: C₇H₁₁F₃O
MW: 168.16 g/mol
Boiling Pt: 70…75 °C/15 mm
Storage Temperature: Ambient
MDL Number: MFCD00102144
CAS Number: 30129-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR