4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine 96%

Supplier: Thermo Scientific Chemicals
Warning

H33260-03 H33260-06 H33260-MD
AAH33260-03EA 102 USD
AAH33260-03 AAH33260-06 AAH33260-MD
4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine 96%
4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine

Purity: 96% CAS No.: 331761-46-7 Molecular Formula: C12H6Cl2N2S Formula Weight: 281.16 Melting Point: 139.5-141° MDL No.: MFCD01038375

Formula: C₁₂H₆Cl₂N₂S
MW: 281.16 g/mol
Melting Pt: 139.5…141 °C
Storage Temperature: Ambient
MDL Number: MFCD01038375
CAS Number: 331761-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR