5-Bromoimidazo[1,2-a]pyridine 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33240-03
H33240-09
AAH33240-03EA
64.6
USD
AAH33240-03
AAH33240-09
5-Bromoimidazo[1,2-a]pyridine 97+%
5-Bromoimidazo[1,2-a]pyridine
Purity: 97+%
CAS No.: 69214-09-1
Molecular Formula: C7H5BrN2
Formula Weight: 197.03
Melting Point: 61-64°
MDL No.: MFCD08275698
Formula:
C₇H₅BrN₂ MW: 197.03 g/mol Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD08275698 CAS Number: 69214-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...