5-Acetyl-2-bromopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Acetyl-6-bromopyridine

H33204-03 H33204-06
AAH33204-03EA 74.6 USD
AAH33204-03 AAH33204-06
5-Acetyl-2-bromopyridine 97%
5-Acetyl-2-bromopyridine

Purity: 97% CAS No.: 139042-59-4 Molecular Formula: C7H6BrNO Formula Weight: 200.03 Melting Point: 124-128° MDL No.: MFCD04974527

Formula: C₇H₆BrNO
MW: 200.03 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD04974527
CAS Number: 139042-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR