4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta[4,5]thieno[2,3-d]pyrimidine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33184-03
H33184-06
AAH33184-03EA
63.6
USD
AAH33184-03
AAH33184-06
AAH33184-MD
4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta[4,5]thieno[2,3-d]pyrimidine 96%
4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta[4,5]thieno[2,3-d]pyrimidine
Purity: 96%
CAS No.: 40106-58-9
Molecular Formula: C11H11ClN2S
Formula Weight: 238.74
Melting Point: 64-66°
MDL No.: MFCD05865130
Formula:
C₁₁H₁₁ClN₂S Melting Pt: 64…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD05865130 CAS Number: 40106-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...