2-Chloro-4,6-diphenyl-1,3,5-triazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H33175-03 H33175-06
AAH33175-03EA 33.5 USD
AAH33175-03 AAH33175-06
2-Chloro-4,6-diphenyl-1,3,5-triazine 97%
2-Chloro-4,6-diphenyl-1,3,5-triazine

Purity: 97% CAS No.: 3842-55-5 Molecular Formula: C15H10ClN3 Formula Weight: 267.72 Melting Point: 138-140° MDL No.: MFCD05738885

Formula: C₁₅H₁₀ClN₃
MW: 267.72 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD05738885
CAS Number: 3842-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR