4,6-Dichloro-2-(trifluoromethyl)pyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

H33142-03 H33142-MD
AAH33142-03EA 82.4 USD
AAH33142-03 AAH33142-MD
4,6-Dichloro-2-(trifluoromethyl)pyrimidine 97%
4,6-Dichloro-2-(trifluoromethyl)pyrimidine

Purity: 97% CAS No.: 705-24-8 Molecular Formula: C5HCl2F3N2 Formula Weight: 216.98 Boiling Point: 50-52°/5mm MDL No.: MFCD08436597

Formula: C₅HCl₂F₃N₂
MW: 216.98 g/mol
Boiling Pt: 50…52 °C/5 mm
Storage Temperature: Ambient
MDL Number: MFCD08436597
CAS Number: 705-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR