6-Chloro-2-iodo-3-hydroxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-2-iodopyridin-3-ol

H33127-03 H33127-09
AAH33127-03EA 71.7 USD
AAH33127-03 AAH33127-09
6-Chloro-2-iodo-3-hydroxypyridine 98%
6-Chloro-2-iodo-3-hydroxypyridine

Purity: 98% CAS No.: 188057-26-3 Molecular Formula: C5H3ClIO Formula Weight: 255.44 Melting Point: 191-193° MDL No.: MFCD07636749

Formula: C₅H₃ClIO
MW: 255.44 g/mol
Melting Pt: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD07636749
CAS Number: 188057-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR