3-Pyrrol-1-ylbenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(1H-Pyrrol-1-yl)benzoic acid

H33124-03 H33124-09
AAH33124-03EA 103 USD
AAH33124-03 AAH33124-09
3-Pyrrol-1-ylbenzoic acid 97%
3-Pyrrol-1-ylbenzoic acid

Purity: 97% CAS No.: 61471-45-2 Molecular Formula: C11H9NO2 Formula Weight: 187.20 Melting Point: 175-178° MDL No.: MFCD02656610

Formula: C₁₁H₉NO₂
MW: 187.2 g/mol
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD02656610
CAS Number: 61471-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR