1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-(2,4-Difluorobenzoyl)piperidin-1-yl)ethanone

H33058-03 H33058-06 H33058-MD
AAH33058-03EA 82 USD
AAH33058-03 AAH33058-06 AAH33058-MD
1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine 96%
1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine

Purity: 96% CAS No.: 84162-82-3 Molecular Formula: C14H15F2NO2 Formula Weight: 267.28 Melting Point: 97-98° MDL No.: MFCD00179440

Formula: C₁₄H₁₅F₂NO₂
MW: 267.28 g/mol
Melting Pt: 97…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00179440
CAS Number: 84162-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR