(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2-Difluoro-1-methylcyclopropanecarboxylic acid
H33055-03
H33055-MD
AAH33055-03EA
192
USD
AAH33055-03
AAH33055-MD
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid 97%
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid
Purity: 97%
CAS No.: 128073-33-6
Molecular Formula: C5H6F2O2
Formula Weight: 136.10
Melting Point: 62-64°
MDL No.: MFCD07777163
Formula:
C₅H₆F₂O₂ Melting Pt: 62…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD07777163 CAS Number: 128073-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...