5-Fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33044-03
H33044-06
AAH33044-03EA
104
USD
AAH33044-03
AAH33044-06
5-Fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride 96%
5-Fluoro-3-piperidin-4-yl-1,2-benzisoxazole hydrochloride
Purity: 96%
CAS No.: 84163-16-6
Molecular Formula: C12H13FN2O.HCl
Formula Weight: 256.71
Melting Point: ca 298° dec.
MDL No.: MFCD06200879
Formula:
C₁₂H₁₄ClFN₂O MW: 256.71 g/mol Melting Pt: ∼ 298 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD06200879 CAS Number: 84163-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...