2-tert-Butyltoluene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33036-03
H33036-06
H33036-MD
AAH33036-03EA
256
USD
AAH33036-03
AAH33036-06
AAH33036-MD
2-tert-Butyltoluene 99%
2-tert-Butyltoluene
Purity: 99%
CAS No.: 1074-92-6
Molecular Formula: C11H16
Formula Weight: 148.25
Refractive Index: 1.5075
Formula:
C₁₁H₁₆ MW: 148.25 g/mol Density: 0.89 Storage Temperature: Ambient |
MDL Number:
MFCD00059209 CAS Number: 1074-92-6 UN: 2667 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...