5-Hydroxynicotinonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H33006-03 H33006-MD
AAH33006-03EA 238 USD
AAH33006-03 AAH33006-MD
5-Hydroxynicotinonitrile 97%
5-Hydroxynicotinonitrile

Purity: 97% CAS No.: 152803-24-2 Molecular Formula: C6H4N2O Formula Weight: 120.11 Melting Point: 213-217° UN No.: 3439 MDL No.: MFCD11044307

Formula: C6H4N2O
MW: 120.11 g/mol
Melting Pt: 213…217 °C
Storage Temperature: Ambient
MDL Number: MFCD11044307
CAS Number: 152803-24-2
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR