2-Bromo-4-(trifluoromethyl)benzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-4-(trifluoromethyl)benzoic acid, 2-Bromo-4-(trifluoromethl)benzoic acid

H32989-03 H32989-06
AAH32989-03EA 125 USD
AAH32989-03 AAH32989-06
2-Bromo-4-(trifluoromethyl)benzoic acid 98%
2-Bromo-4-(trifluoromethyl)benzoic acid

Purity: 98% CAS No.: 328-89-2 Molecular Formula: C8H4BrF3O2 Formula Weight: 269.02 MDL No.: MFCD07368837

Formula: C₈H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07368837
CAS Number: 328-89-2
EINECS: 206-339-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR