4-Bromo-2,3,5,6-tetrafluorobenzoic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

H32982-03 H32982-06
AAH32982-03EA 103 USD
AAH32982-03 AAH32982-06
4-Bromo-2,3,5,6-tetrafluorobenzoic acid 97+%
4-Bromo-2,3,5,6-tetrafluorobenzoic acid

Purity: 97+% CAS No.: 4707-24-8 Molecular Formula: C7HBrF4O2 Formula Weight: 272.98 Odor: Odorless Melting Point: 142-145° MDL No.: MFCD00075244

Formula: C₇HBrF₄O₂
MW: 272.98
Melting Pt: 142…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00075244
CAS Number: 4707-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR