Furo[3,2-b]pyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H32966-06
H32966-03
AAH32966-06EA
783
USD
AAH32966-06
AAH32966-MD
AAH32966-03
Furo[3,2-b]pyridine 98%
Furo[3,2-b]pyridine
Purity: 98%
CAS No.: 272-62-8
Molecular Formula: C7H5NO
Formula Weight: 119.12
Refractive Index: 1.5753
Boiling Point: 79-80°/12mm
MDL No.: MFCD11870744
Formula:
C₇H₅NO MW: 119.12 g/mol Boiling Pt: 79…80 °C/12 mm Storage Temperature: Ambient |
MDL Number:
MFCD11870744 CAS Number: 272-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...