1-Bromo-4-dodecylbenzene 98%

Supplier: Thermo Scientific Chemicals

H32963-06 H32963-14
AAH32963-03EA 65.52 USD
AAH32963-03 AAH32963-06 AAH32963-14
1-Bromo-4-dodecylbenzene 98%
1-Bromo-4-dodecylbenzene

Purity: 98% CAS No.: 126930-72-1 Molecular Formula: C18H29Br Formula Weight: 325.33 Flash Point: 110°(230°F) Refractive Index: 1.5034 Boiling Point: 376° MDL No.: MFCD00191386

Formula: C₁₈H₂₉Br
MW: 325.33 g/mol
Boiling Pt: 376 °C
Density: 1.07
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191386
CAS Number: 126930-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR